Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTTAATAAGGTGCACAAATATGC[A/T]GATATCCTGTATTATGAGCATGTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614136 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRAPPC8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
TRAPPC8 - trafficking protein particle complex 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014939.3 | 4855 | UTR 3 | NP_055754.2 | |||
XM_005258233.2 | 4855 | UTR 3 | XP_005258290.1 | |||
XM_005258234.2 | 4855 | UTR 3 | XP_005258291.1 | |||
XM_005258235.3 | 4855 | UTR 3 | XP_005258292.1 | |||
XM_006722420.3 | 4855 | Intron | XP_006722483.1 | |||
XM_011525884.2 | 4855 | UTR 3 | XP_011524186.1 | |||
XM_017025615.1 | 4855 | UTR 3 | XP_016881104.1 | |||
XM_017025616.1 | 4855 | UTR 3 | XP_016881105.1 | |||
XM_017025617.1 | 4855 | UTR 3 | XP_016881106.1 |