Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAAATGAGTATAATTTTTTTCTC[C/T]CTCAAAATGTAATTTTATCTGGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300317 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
REPS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
REPS2 - RALBP1 associated Eps domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080975.1 | Intron | NP_001074444.1 | ||||
NM_004726.2 | Intron | NP_004717.2 | ||||
XM_005274625.3 | Intron | XP_005274682.1 | ||||
XM_005274626.3 | Intron | XP_005274683.1 | ||||
XM_011545603.2 | Intron | XP_011543905.1 | ||||
XM_011545604.2 | Intron | XP_011543906.1 | ||||
XM_011545605.2 | Intron | XP_011543907.1 | ||||
XM_011545606.2 | Intron | XP_011543908.1 | ||||
XM_011545607.2 | Intron | XP_011543909.1 | ||||
XM_011545609.2 | Intron | XP_011543911.1 | ||||
XM_017029955.1 | Intron | XP_016885444.1 | ||||
XM_017029956.1 | Intron | XP_016885445.1 | ||||
XM_017029957.1 | Intron | XP_016885446.1 | ||||
XM_017029958.1 | Intron | XP_016885447.1 |
Set Membership: |
HapMap |