Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCACCTTTAATTTGACTTATGATT[A/G]GGGCAGGCCGACTGAACAAACTCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TPM3P9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TPM3P9 - tropomyosin 3 pseudogene 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF761 - zinc finger protein 761 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008401.3 | Intron | NP_001008401.3 | ||||
NM_001289951.1 | Intron | NP_001276880.1 | ||||
NM_001289952.1 | Intron | NP_001276881.1 | ||||
NM_001289953.1 | Intron | NP_001276882.1 |
Set Membership: |
HapMap |