Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGAAGTGAAATTACTCCATCAAAC[C/T]CATTCCTGGATTCTCTTCTAGAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611957 MIM: 608886 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR378A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MIR378A - microRNA 378a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPARGC1B - PPARG coactivator 1 beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172698.1 | Intron | NP_001166169.1 | ||||
NM_001172699.1 | Intron | NP_001166170.1 | ||||
NM_133263.3 | Intron | NP_573570.3 | ||||
XM_005268372.4 | Intron | XP_005268429.1 | ||||
XM_011537553.2 | Intron | XP_011535855.1 | ||||
XM_011537554.2 | Intron | XP_011535856.1 | ||||
XM_011537555.2 | Intron | XP_011535857.1 | ||||
XM_011537556.2 | Intron | XP_011535858.1 | ||||
XM_011537557.1 | Intron | XP_011535859.1 |
Set Membership: |
HapMap |