Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCTCTCTGTTCCCAGCTTTTCTG[C/T]ACTTCCTCTGCTCTCCAGCTCCAAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605350 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL27RA PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IL27RA - interleukin 27 receptor subunit alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PALM3 - paralemmin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145028.1 | 1318 | Missense Mutation | ACA,GCA | T,A 440 | NP_001138500.1 | |
XM_005259891.4 | 1318 | Missense Mutation | ACA,GCA | T,A 455 | XP_005259948.1 | |
XM_011527965.2 | 1318 | Missense Mutation | ACA,GCA | T,A 428 | XP_011526267.1 |