Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTGCCTCATGGGTTTGGCTGCCC[C/T]GTTTTGAAGTTACCCAAAGATTATT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147730 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL2RA PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IL2RA - interleukin 2 receptor subunit alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000417.2 | 1715 | UTR 3 | NP_000408.1 | |||
NM_001308242.1 | 1715 | UTR 3 | NP_001295171.1 | |||
NM_001308243.1 | 1715 | UTR 3 | NP_001295172.1 |
Set Membership: |
HapMap |