Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCCTGAGTGATGTTCCCAGCACC[A/G]TTTAAATGTTCTACATGGGTTATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604603 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYOF PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MYOF - myoferlin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013451.3 | Intron | NP_038479.1 | ||||
NM_133337.2 | Intron | NP_579899.1 | ||||
XM_005269693.4 | Intron | XP_005269750.1 | ||||
XM_005269694.4 | Intron | XP_005269751.1 | ||||
XM_011539632.2 | Intron | XP_011537934.1 | ||||
XM_017016068.1 | Intron | XP_016871557.1 | ||||
XM_017016069.1 | Intron | XP_016871558.1 | ||||
XM_017016070.1 | Intron | XP_016871559.1 |
Set Membership: |
HapMap |