Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGCTCTGTGAATATAGGTAAGT[A/C]TCTTCTCCTTGTTTTAGTTTCCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602883 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLCO1A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SLCO1A2 - solute carrier organic anion transporter family member 1A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021094.3 | 4034 | UTR 3 | NP_066580.1 | |||
NM_134431.3 | 4034 | UTR 3 | NP_602307.1 | |||
XM_005253474.3 | 4034 | UTR 3 | XP_005253531.1 | |||
XM_005253477.3 | 4034 | UTR 3 | XP_005253534.1 | |||
XM_011520818.1 | 4034 | UTR 3 | XP_011519120.1 | |||
XM_011520819.1 | 4034 | UTR 3 | XP_011519121.1 | |||
XM_011520820.2 | 4034 | UTR 3 | XP_011519122.1 | |||
XM_011520821.2 | 4034 | UTR 3 | XP_011519123.1 | |||
XM_017019849.1 | 4034 | Intron | XP_016875338.1 | |||
XM_017019850.1 | 4034 | Intron | XP_016875339.1 |
Set Membership: |
HapMap |