Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACGTGGAGACTTGATGCATTTTTG[A/C]TGTGGACGAAAGGGCCCGGGGGCGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605032 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPLX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CPLX1 - complexin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006651.3 | 1603 | UTR 3 | NP_006642.1 | |||
XM_011513391.1 | 1603 | UTR 3 | XP_011511693.1 |
LOC100129917 - uncharacterized LOC100129917 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |