Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607766 MIM: 614552 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACAP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ACAP2 - ArfGAP with coiled-coil, ankyrin repeat and PH domains 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012287.5 | 2765 | UTR 3 | NP_036419.3 | |||
XM_006713557.3 | 2765 | UTR 3 | XP_006713620.1 | |||
XM_011512602.2 | 2765 | UTR 3 | XP_011510904.1 | |||
XM_011512603.2 | 2765 | UTR 3 | XP_011510905.1 | |||
XM_011512604.2 | 2765 | UTR 3 | XP_011510906.1 | |||
XM_011512605.2 | 2765 | UTR 3 | XP_011510907.1 | |||
XM_011512606.2 | 2765 | UTR 3 | XP_011510908.1 | |||
XM_017006046.1 | 2765 | UTR 3 | XP_016861535.1 | |||
XM_017006047.1 | 2765 | UTR 3 | XP_016861536.1 | |||
XM_017006048.1 | 2765 | UTR 3 | XP_016861537.1 | |||
XM_017006049.1 | 2765 | UTR 3 | XP_016861538.1 |
XXYLT1 - xyloside xylosyltransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |