Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGACTAACTTGGATACGAAAGTT[C/T]AGAATAATGCCCACCAAAAATCCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612116 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105369205 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LOC105369205 - uncharacterized LOC105369205 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011524128.2 | 4350 | Intron | XP_011522430.1 | |||
XM_011524129.2 | 4350 | Intron | XP_011522431.1 | |||
XM_011524131.2 | 4350 | UTR 3 | XP_011522433.1 | |||
XM_017025507.1 | 4350 | Intron | XP_016880996.1 | |||
XM_017025508.1 | 4350 | Intron | XP_016880997.1 | |||
XM_017025509.1 | 4350 | Intron | XP_016880998.1 |
USP22 - ubiquitin specific peptidase 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015276.1 | 4350 | UTR 3 | NP_056091.1 | |||
XM_005256575.2 | 4350 | UTR 3 | XP_005256632.1 |
Set Membership: |
HapMap Validated |