Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602654 MIM: 604984 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FARP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
FARP1 - FERM, ARH/RhoGEF and pleckstrin domain protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STK24 - serine/threonine kinase 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032296.3 | 3084 | UTR 3 | NP_001027467.2 | |||
NM_001286649.1 | 3084 | UTR 3 | NP_001273578.1 | |||
NM_003576.4 | 3084 | UTR 3 | NP_003567.2 | |||
XM_005254079.4 | 3084 | Intron | XP_005254136.1 | |||
XM_017020794.1 | 3084 | Intron | XP_016876283.1 | |||
XM_017020795.1 | 3084 | Intron | XP_016876284.1 |