Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACCCATCCATTTATTCAGCAAAC[A/G]TTGGCAAGTAATATGCCTGGAGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614930 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LRRC6 - leucine rich repeat containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321961.1 | Intron | NP_001308890.1 | ||||
NM_001321962.1 | Intron | NP_001308891.1 | ||||
NM_001321963.1 | Intron | NP_001308892.1 | ||||
NM_001321964.1 | Intron | NP_001308893.1 | ||||
NM_001321965.1 | Intron | NP_001308894.1 | ||||
NM_001321966.1 | Intron | NP_001308895.1 | ||||
NM_012472.5 | Intron | NP_036604.2 | ||||
XM_006716538.3 | Intron | XP_006716601.2 | ||||
XM_011516950.2 | Intron | XP_011515252.1 | ||||
XM_017013296.1 | Intron | XP_016868785.1 | ||||
XM_017013297.1 | Intron | XP_016868786.1 | ||||
XM_017013298.1 | Intron | XP_016868787.1 |