Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCACACCAAAGTACCCAAGGGAGC[A/G]GACTCAGTAAAAGGGATTATCAGGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
41 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 153390 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM167B PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
FAM167B - family with sequence similarity 167 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LCK - LCK proto-oncogene, Src family tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042771.2 | Intron | NP_001036236.1 | ||||
NM_005356.4 | Intron | NP_005347.3 | ||||
XM_011541453.2 | Intron | XP_011539755.1 | ||||
XM_011541454.2 | Intron | XP_011539756.1 | ||||
XM_011541455.2 | Intron | XP_011539757.1 |