Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAGAATTGATGGAGATGTCTGAT[A/G]TATCAGAGGCATTTAATAAATAGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
17 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607410 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DMBX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DMBX1 - diencephalon/mesencephalon homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_147192.2 | Intron | NP_671725.1 | ||||
NM_172225.1 | Intron | NP_757379.1 | ||||
XM_011540668.2 | Intron | XP_011538970.1 | ||||
XM_017000289.1 | Intron | XP_016855778.1 |
Set Membership: |
HapMap |