Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603295 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMAD9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SMAD9 - SMAD family member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127217.2 | 2251 | UTR 3 | NP_001120689.1 | |||
NM_005905.5 | 2251 | UTR 3 | NP_005896.1 | |||
XM_005266401.3 | 2251 | Intron | XP_005266458.1 | |||
XM_005266403.3 | 2251 | Intron | XP_005266460.1 | |||
XM_005266404.3 | 2251 | Intron | XP_005266461.1 | |||
XM_006719827.3 | 2251 | Intron | XP_006719890.1 |
Set Membership: |
HapMap |