Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCCAAGTGACTTAACCAGGTCGG[C/T]TGGACATATCCCACACCTAGAAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 186982 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TCP11 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TCP11 - t-complex 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001093728.2 | Intron | NP_001087197.1 | ||||
NM_001261817.1 | Intron | NP_001248746.1 | ||||
NM_001261818.1 | Intron | NP_001248747.1 | ||||
NM_001261819.1 | Intron | NP_001248748.1 | ||||
NM_001261820.1 | Intron | NP_001248749.1 | ||||
NM_001261821.1 | Intron | NP_001248750.1 | ||||
NM_018679.5 | Intron | NP_061149.1 | ||||
XM_005249339.2 | Intron | XP_005249396.1 | ||||
XM_011514830.1 | Intron | XP_011513132.1 | ||||
XM_011514831.1 | Intron | XP_011513133.1 | ||||
XM_011514832.1 | Intron | XP_011513134.1 |
Set Membership: |
HapMap |