Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607942 MIM: 616872 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIK3AP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PIK3AP1 - phosphoinositide-3-kinase adaptor protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152309.2 | 4006 | UTR 3 | NP_689522.2 | |||
XM_005269498.1 | 4006 | UTR 3 | XP_005269555.1 | |||
XM_005269499.1 | 4006 | UTR 3 | XP_005269556.1 | |||
XM_011539248.1 | 4006 | UTR 3 | XP_011537550.1 | |||
XM_011539249.1 | 4006 | UTR 3 | XP_011537551.1 |
TM9SF3 - transmembrane 9 superfamily member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |