Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607942 MIM: 616872 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIK3AP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PIK3AP1 - phosphoinositide-3-kinase adaptor protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152309.2 | 3054 | UTR 3 | NP_689522.2 | |||
XM_005269498.1 | 3054 | UTR 3 | XP_005269555.1 | |||
XM_005269499.1 | 3054 | UTR 3 | XP_005269556.1 | |||
XM_011539248.1 | 3054 | UTR 3 | XP_011537550.1 | |||
XM_011539249.1 | 3054 | UTR 3 | XP_011537551.1 |
TM9SF3 - transmembrane 9 superfamily member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |