Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 126065 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP24A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CYP24A1 - cytochrome P450 family 24 subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000782.4 | 2550 | Intron | NP_000773.2 | |||
NM_001128915.1 | 2550 | Intron | NP_001122387.1 | |||
XM_005260304.4 | 2550 | Intron | XP_005260361.1 | |||
XM_017027691.1 | 2550 | Intron | XP_016883180.1 | |||
XM_017027692.1 | 2550 | UTR 3 | XP_016883181.1 | |||
XM_017027693.1 | 2550 | Intron | XP_016883182.1 |
Set Membership: |
HapMap |