Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616635 MIM: 604546 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYHR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYHR1 - cysteine and histidine rich 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001129888.1 | 3076 | Intron | NP_001123360.1 | |||
NM_032687.3 | 3076 | Intron | NP_116076.1 | |||
NM_138496.1 | 3076 | Intron | NP_612505.1 | |||
XM_006716560.3 | 3076 | Intron | XP_006716623.1 | |||
XM_006716561.3 | 3076 | Intron | XP_006716624.1 | |||
XM_006716562.3 | 3076 | Intron | XP_006716625.2 | |||
XM_011517051.1 | 3076 | UTR 5 | XP_011515353.1 | |||
XM_017013463.1 | 3076 | Intron | XP_016868952.1 | |||
XM_017013464.1 | 3076 | Intron | XP_016868953.1 | |||
XM_017013465.1 | 3076 | Intron | XP_016868954.1 | |||
XM_017013466.1 | 3076 | Intron | XP_016868955.1 |
TONSL - tonsoku-like, DNA repair protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |