Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142290 MIM: 605493 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HPX PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
HPX - hemopexin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRIM3 - tripartite motif containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001248006.1 | 2612 | UTR 3 | NP_001234935.1 | |||
NM_001248007.1 | 2612 | UTR 3 | NP_001234936.1 | |||
NM_006458.3 | 2612 | UTR 3 | NP_006449.2 | |||
NM_033278.3 | 2612 | UTR 3 | NP_150594.2 | |||
XM_011519844.1 | 2612 | UTR 3 | XP_011518146.1 | |||
XM_011519845.2 | 2612 | UTR 3 | XP_011518147.1 | |||
XM_017017098.1 | 2612 | UTR 3 | XP_016872587.1 | |||
XM_017017099.1 | 2612 | UTR 3 | XP_016872588.1 |
Set Membership: |
HapMap Validated |