Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGATTTCTTTGAAATAGGCAGTAG[A/C]GACAATTTTAAAGTACTTAGTTGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603566 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIAS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PIAS1 - protein inhibitor of activated STAT 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320687.1 | 257 | Intron | NP_001307616.1 | |||
NM_016166.2 | 257 | Intron | NP_057250.1 | |||
XM_011522126.2 | 257 | Intron | XP_011520428.1 | |||
XM_011522127.2 | 257 | Intron | XP_011520429.1 | |||
XM_017022688.1 | 257 | Intron | XP_016878177.1 | |||
XM_017022689.1 | 257 | Intron | XP_016878178.1 | |||
XM_017022690.1 | 257 | UTR 5 | XP_016878179.1 | |||
XM_017022691.1 | 257 | Intron | XP_016878180.1 |