Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610124 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C3orf22 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C3orf22 - chromosome 3 open reading frame 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152533.2 | 1683 | Intron | NP_689746.1 | |||
XM_011512455.2 | 1683 | Intron | XP_011510757.1 | |||
XM_011512457.2 | 1683 | Intron | XP_011510759.1 | |||
XM_017005751.1 | 1683 | Intron | XP_016861240.1 | |||
XM_017005752.1 | 1683 | Intron | XP_016861241.1 | |||
XM_017005753.1 | 1683 | UTR 3 | XP_016861242.1 |
CHST13 - carbohydrate sulfotransferase 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152889.2 | 1683 | Intron | NP_690849.1 |
Set Membership: |
HapMap |