Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTTGGCCACAGAACCCTCTATTC[A/T]CCTAACAATATATTCTGACAAAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTX3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MTX3 - metaxin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010891.4 | 5713 | UTR 3 | NP_001010891.4 | |||
NM_001167741.1 | 5713 | UTR 3 | NP_001161213.1 | |||
XM_005248495.3 | 5713 | UTR 3 | XP_005248552.1 | |||
XM_017009440.1 | 5713 | UTR 3 | XP_016864929.1 |