Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTTCCTCCTCCTCTAAGCAGGC[A/G]GCAGGGGAAGGTGGAGGGGTTGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 167415 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAX8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PAX8 - paired box 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003466.3 | Intron | NP_003457.1 | ||||
NM_013952.3 | Intron | NP_039246.1 | ||||
NM_013953.3 | Intron | NP_039247.1 | ||||
NM_013992.3 | Intron | NP_054698.1 | ||||
XM_011511790.1 | Intron | XP_011510092.1 | ||||
XM_011511791.1 | Intron | XP_011510093.1 | ||||
XM_011511792.1 | Intron | XP_011510094.1 | ||||
XM_011511793.1 | Intron | XP_011510095.1 | ||||
XM_011511794.1 | Intron | XP_011510096.1 | ||||
XM_017004878.1 | Intron | XP_016860367.1 | ||||
XM_017004879.1 | Intron | XP_016860368.1 |
PAX8-AS1 - PAX8 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |