Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACTGTAGAGTTAGTATGCACTGC[A/G]ATCCCAAACATTTTCTTGTTCCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611128 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MDGA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MDGA2 - MAM domain containing glycosylphosphatidylinositol anchor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113498.2 | 3628 | UTR 3 | NP_001106970.3 | |||
NM_182830.4 | 3628 | UTR 3 | NP_878250.2 | |||
XM_011536519.2 | 3628 | UTR 3 | XP_011534821.1 | |||
XM_011536522.2 | 3628 | Intron | XP_011534824.1 | |||
XM_011536525.2 | 3628 | UTR 3 | XP_011534827.1 | |||
XM_017021060.1 | 3628 | UTR 3 | XP_016876549.1 | |||
XM_017021061.1 | 3628 | Intron | XP_016876550.1 |