Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606365 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GLS2 - glutaminase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001280796.1 | 2206 | UTR 3 | NP_001267725.1 | |||
NM_001280797.1 | 2206 | UTR 3 | NP_001267726.1 | |||
NM_001280798.1 | 2206 | UTR 3 | NP_001267727.1 | |||
NM_013267.3 | 2206 | UTR 3 | NP_037399.2 | |||
XM_005268797.1 | 2206 | UTR 3 | XP_005268854.1 | |||
XM_017019180.1 | 2206 | UTR 3 | XP_016874669.1 |
SPRYD4 - SPRY domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207344.3 | 2206 | Intron | NP_997227.1 |
Set Membership: |
HapMap |