Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTTTTACTTTGCTGGGATGTTTC[C/G]CATCACCAACATTGAGTTTCTTCCC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
TMPRSS7 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
TMPRSS7 - transmembrane protease, serine 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042575.2 | 581 | Missense Mutation | CCC,CGC | P,R 3 | NP_001036040.2 | |
XM_011512754.1 | 581 | Missense Mutation | CCC,CGC | P,R 32 | XP_011511056.1 | |
XM_011512758.1 | 581 | Intron | XP_011511060.1 |