Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
59 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616877 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TMEM9 - transmembrane protein 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288564.1 | Intron | NP_001275493.1 | ||||
NM_001288565.1 | Intron | NP_001275494.1 | ||||
NM_001288566.1 | Intron | NP_001275495.1 | ||||
NM_001288567.1 | Intron | NP_001275496.1 | ||||
NM_001288568.1 | Intron | NP_001275497.1 | ||||
NM_001288569.1 | Intron | NP_001275498.1 | ||||
NM_001288570.1 | Intron | NP_001275499.1 | ||||
NM_001288571.1 | Intron | NP_001275500.1 | ||||
NM_016456.4 | Intron | NP_057540.1 | ||||
XM_011509385.1 | Intron | XP_011507687.1 | ||||
XM_011509386.2 | Intron | XP_011507688.1 | ||||
XM_011509387.2 | Intron | XP_011507689.1 |