Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DZIP1L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DZIP1L - DAZ interacting zinc finger protein 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170538.1 | Intron | NP_001164009.1 | ||||
NM_173543.2 | Intron | NP_775814.2 | ||||
XM_005247198.3 | Intron | XP_005247255.1 | ||||
XM_005247200.4 | Intron | XP_005247257.1 | ||||
XM_006713526.1 | Intron | XP_006713589.1 | ||||
XM_006713527.3 | Intron | XP_006713590.1 | ||||
XM_011512518.2 | Intron | XP_011510820.1 | ||||
XM_011512520.2 | Intron | XP_011510822.1 | ||||
XM_011512521.2 | Intron | XP_011510823.1 | ||||
XM_011512522.1 | Intron | XP_011510824.1 | ||||
XM_011512524.2 | Intron | XP_011510826.1 | ||||
XM_011512528.2 | Intron | XP_011510830.1 | ||||
XM_017005841.1 | Intron | XP_016861330.1 | ||||
XM_017005842.1 | Intron | XP_016861331.1 |