Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
67 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
USP48 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
USP48 - ubiquitin specific peptidase 48 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032730.1 | Intron | NP_001027902.1 | ||||
XM_005246009.2 | Intron | XP_005246066.1 | ||||
XM_006710951.2 | Intron | XP_006711014.2 | ||||
XM_006710955.3 | Intron | XP_006711018.1 | ||||
XM_011542262.2 | Intron | XP_011540564.1 | ||||
XM_011542263.2 | Intron | XP_011540565.1 | ||||
XM_011542264.1 | Intron | XP_011540566.1 | ||||
XM_011542265.2 | Intron | XP_011540567.1 | ||||
XM_011542267.2 | Intron | XP_011540569.1 | ||||
XM_011542268.2 | Intron | XP_011540570.1 | ||||
XM_017002495.1 | Intron | XP_016857984.1 | ||||
XM_017002496.1 | Intron | XP_016857985.1 | ||||
XM_017002497.1 | Intron | XP_016857986.1 |
Set Membership: |
HapMap |