Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
58 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600114 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CCT3 - chaperonin containing TCP1 subunit 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSACC - TSSK6 activating cochaperone | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304817.1 | Intron | NP_001291746.1 | ||||
NM_001304818.1 | Intron | NP_001291747.1 | ||||
NM_001304819.1 | Intron | NP_001291748.1 | ||||
NM_001304820.1 | Intron | NP_001291749.1 | ||||
NM_001304821.1 | Intron | NP_001291750.1 | ||||
NM_001304822.1 | Intron | NP_001291751.1 | ||||
NM_001304823.1 | Intron | NP_001291752.1 | ||||
NM_001304824.1 | Intron | NP_001291753.1 | ||||
NM_001304825.1 | Intron | NP_001291754.1 | ||||
NM_001304826.1 | Intron | NP_001291755.1 | ||||
NM_144627.4 | Intron | NP_653228.1 |
Set Membership: |
HapMap JSNP Validated |