Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTAAGGGCCGGCTCATCCAGGGA[C/T]GGTGACTACCTACAAACTAGGGCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602146 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LHX4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LHX4 - LIM homeobox 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033343.3 | Intron | NP_203129.1 | ||||
XM_011510105.2 | Intron | XP_011508407.1 | ||||
XM_011510106.2 | Intron | XP_011508408.1 | ||||
XM_011510108.2 | Intron | XP_011508410.1 | ||||
XM_017002755.1 | Intron | XP_016858244.1 |