Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTTACTTTTTTTTTTCTTTTTG[G/T]TACTTGTACTGTGGTAAGAAAAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 616751 MIM: 613360 | ||||||||||||||||||||
Literature Links: |
CEPT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CEPT1 - choline/ethanolamine phosphotransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_006710285.3 | Intron | XP_006710348.1 | ||||
XM_011540496.1 | Intron | XP_011538798.1 |
DRAM2 - DNA damage regulated autophagy modulator 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |