Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAATCGGGGAGGGCAATCAGGGCA[A/G]TCATCTACTGTGGAGTAGAAGGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606646 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARAP1 - ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040118.2 | Intron | NP_001035207.1 | ||||
NM_001135190.1 | Intron | NP_001128662.1 | ||||
NM_015242.4 | Intron | NP_056057.2 |
Set Membership: |
HapMap |