Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCAGCCTCACCCGCCCTGTGGCT[A/G]ACCCCCAAGCCCCATGCTAGATCAC
Species: |
Human | |||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606775 | |||||||||||||||||||||||||||||||||||
Literature Links: |
CSPG5 PubMed Links | |||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | ||||||
SAS - Not Available | Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR - Not Available | Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR - Not Available | ||||||||
AMR - Not Available |
CSPG5 - chondroitin sulfate proteoglycan 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206942.1 | Intron | NP_001193871.1 | ||||
NM_001206943.1 | Intron | NP_001193872.1 | ||||
NM_001206944.1 | Intron | NP_001193873.1 | ||||
NM_001206945.1 | Intron | NP_001193874.1 | ||||
NM_006574.3 | Intron | NP_006565.2 |