Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107323 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALDH7A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ALDH7A1 - aldehyde dehydrogenase 7 family member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001182.4 | 4834 | UTR 3 | NP_001173.2 | |||
NM_001201377.1 | 4834 | UTR 3 | NP_001188306.1 | |||
NM_001202404.1 | 4834 | UTR 3 | NP_001189333.2 | |||
XM_011543417.2 | 4834 | Intron | XP_011541719.1 | |||
XM_017009493.1 | 4834 | Intron | XP_016864982.1 |
Set Membership: |
HapMap |