Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTCTTAATGGCTTTTAATATAAA[C/T]ATGCATCCCTTCTCCTTTGAAATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609816 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NBEAL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NBEAL1 - neurobeachin like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114132.1 | Intron | NP_001107604.1 | ||||
XM_005246787.3 | Intron | XP_005246844.1 | ||||
XM_005246788.2 | Intron | XP_005246845.1 | ||||
XM_006712698.3 | Intron | XP_006712761.1 | ||||
XM_006712699.3 | Intron | XP_006712762.1 | ||||
XM_011511658.2 | Intron | XP_011509960.1 | ||||
XM_011511659.2 | Intron | XP_011509961.1 | ||||
XM_011511660.2 | Intron | XP_011509962.1 | ||||
XM_011511662.2 | Intron | XP_011509964.1 | ||||
XM_011511663.2 | Intron | XP_011509965.1 | ||||
XM_017004728.1 | Intron | XP_016860217.1 | ||||
XM_017004729.1 | Intron | XP_016860218.1 |