Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCAACAGAGTGGTTTCGTGTTTG[C/T]TTCTGCCATTTACCTCAGGAATATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603725 MIM: 608073 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FGF17 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
FGF17 - fibroblast growth factor 17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NPM2 - nucleophosmin/nucleoplasmin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286680.1 | Intron | NP_001273609.1 | ||||
NM_001286681.1 | Intron | NP_001273610.1 | ||||
NM_182795.1 | Intron | NP_877724.1 | ||||
XM_011544360.2 | Intron | XP_011542662.1 | ||||
XM_011544362.2 | Intron | XP_011542664.1 | ||||
XM_011544363.2 | Intron | XP_011542665.1 | ||||
XM_011544364.2 | Intron | XP_011542666.1 | ||||
XM_017012947.1 | Intron | XP_016868436.1 | ||||
XM_017012948.1 | Intron | XP_016868437.1 | ||||
XM_017012949.1 | Intron | XP_016868438.1 | ||||
XM_017012950.1 | Intron | XP_016868439.1 |
Set Membership: |
HapMap Validated |