Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608295 MIM: 608619 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM107A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM107A - family with sequence similarity 107 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM3D - family with sequence similarity 3 member D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138805.2 | Intron | NP_620160.1 | ||||
XM_005264862.4 | Intron | XP_005264919.1 | ||||
XM_005264863.4 | Intron | XP_005264920.1 | ||||
XM_005264864.4 | Intron | XP_005264921.1 | ||||
XM_006712965.3 | Intron | XP_006713028.1 | ||||
XM_006712967.3 | Intron | XP_006713030.1 | ||||
XM_011533348.2 | Intron | XP_011531650.1 | ||||
XM_011533349.2 | Intron | XP_011531651.1 | ||||
XM_011533350.2 | Intron | XP_011531652.1 | ||||
XM_011533351.2 | Intron | XP_011531653.1 | ||||
XM_011533352.2 | Intron | XP_011531654.1 | ||||
XM_011533353.2 | Intron | XP_011531655.1 | ||||
XM_011533354.2 | Intron | XP_011531656.1 |
FAM3D-AS1 - FAM3D antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |