Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGACCTGTGATTTCTTTATTCATC[C/T]GGGAGCCTTCAGGCCCCTAAAAAAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
46 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600871 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GFI1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GFI1 - growth factor independent 1 transcriptional repressor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127215.1 | 3245 | Intron | NP_001120687.1 | |||
NM_001127216.1 | 3245 | Intron | NP_001120688.1 | |||
NM_005263.3 | 3245 | Intron | NP_005254.2 | |||
XM_005270749.3 | 3245 | UTR 3 | XP_005270806.1 | |||
XM_011541245.2 | 3245 | UTR 3 | XP_011539547.1 | |||
XM_011541246.2 | 3245 | UTR 3 | XP_011539548.1 |
Set Membership: |
HapMap |