Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAGTTTAGCCAAGCAGACAAGTT[G/T]CCATATTGCTAAGGAACAATCACCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114021 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDH3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CDH3 - cadherin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317195.1 | Intron | NP_001304124.1 | ||||
NM_001317196.1 | Intron | NP_001304125.1 | ||||
NM_001793.5 | Intron | NP_001784.2 | ||||
XM_011522800.2 | Intron | XP_011521102.1 |
Set Membership: |
HapMap Validated |