Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605600 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IPO8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IPO8 - importin 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190995.1 | 3710 | UTR 3 | NP_001177924.1 | |||
NM_006390.3 | 3710 | UTR 3 | NP_006381.2 | |||
XM_017018691.1 | 3710 | UTR 3 | XP_016874180.1 | |||
XM_017018692.1 | 3710 | UTR 3 | XP_016874181.1 | |||
XM_017018693.1 | 3710 | Intron | XP_016874182.1 |
Set Membership: |
HapMap |