Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604763 MIM: 607394 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGEF12 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ARHGEF12 - Rho guanine nucleotide exchange factor 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POU2F3 - POU class 2 homeobox 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM136 - transmembrane protein 136 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198670.1 | 503 | Silent Mutation | CAA,CAG | Q,Q 3 | NP_001185599.1 | |
NM_001198671.1 | 503 | Intron | NP_001185600.1 | |||
NM_001198672.1 | 503 | Intron | NP_001185601.1 | |||
NM_001198673.1 | 503 | Intron | NP_001185602.1 | |||
NM_001198674.1 | 503 | Intron | NP_001185603.1 | |||
NM_001198675.1 | 503 | Intron | NP_001185604.1 | |||
NM_174926.2 | 503 | Silent Mutation | CAA,CAG | Q,Q 3 | NP_777586.1 | |
XM_017017327.1 | 503 | Silent Mutation | CAA,CAG | Q,Q 3 | XP_016872816.1 |