Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATACCTGAGACCTAGTAATTGATAC[A/G]GAAAAGGGGTTTGGCTGGGCATGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
52 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611680 MIM: 612023 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf116 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C1orf116 - chromosome 1 open reading frame 116 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
YOD1 - YOD1 deubiquitinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |