Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTATCCAAAACCTTGGATCTTTCA[G/T]CAGTGCTTTATCAAAGCAGCTAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
87 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611923 MIM: 608962 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GJA9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GJA9 - gap junction protein alpha 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GJA9-MYCBP - GJA9-MYCBP readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RHBDL2 - rhomboid like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304746.1 | 1061 | Missense Mutation | ATG,CTG | M,L 353 | NP_001291675.1 | |
NM_017821.4 | 1061 | Missense Mutation | ATG,CTG | M,L 273 | NP_060291.2 |