Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C10orf67 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
C10orf67 - chromosome 10 open reading frame 67 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153714.2 | Intron | NP_714925.2 | ||||
XM_011519440.1 | Intron | XP_011517742.1 | ||||
XM_011519441.1 | Intron | XP_011517743.1 | ||||
XM_011519442.2 | Intron | XP_011517744.1 | ||||
XM_011519443.1 | Intron | XP_011517745.1 | ||||
XM_011519444.2 | Intron | XP_011517746.1 | ||||
XM_011519445.2 | Intron | XP_011517747.1 | ||||
XM_011519446.1 | Intron | XP_011517748.1 | ||||
XM_017016029.1 | Intron | XP_016871518.1 | ||||
XM_017016030.1 | Intron | XP_016871519.1 | ||||
XM_017016031.1 | Intron | XP_016871520.1 |