Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATGACTTTTGCCTGTTTACATTT[A/G]TTCCATCAACCTGAATTTTCCCATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601131 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP2C18 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CYP2C18 - cytochrome P450 family 2 subfamily C member 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000772.2 | Intron | NP_000763.1 | ||||
NM_001128925.1 | Intron | NP_001122397.1 |
Set Membership: |
HapMap Validated |