Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGCATTTCATTCTTGACCTCAGG[A/G]CTCGGACTAGCCTGAGGATCGGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613851 MIM: 616884 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRIMA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRIMA1 - proline rich membrane anchor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178013.3 | 3071 | UTR 3 | NP_821092.1 | |||
XM_011536456.2 | 3071 | UTR 3 | XP_011534758.1 |
UNC79 - unc-79 homolog (C. elegans) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |